Description
- Rounded micro hole interior basket without corners
- Filtration enhanced by side vents
- Two-piece lid design
- Processed biopsies are easy to find
- Various colors available
- Tubed, loose and taped options for packaging
Specifications
Specifications
| Packaging Type | Taped |
| Color | Blue |
| Product Type | Biopsy Micro Cassette |