Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cancer testis antigen 65 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987467
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7467 Supplier Bioss Supplier No. BS6746R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TULP2 is a member of a family of tubby-like genes (TULPs) that encode proteins of unknown function. Members of this family have been identified in plants, vertebrates, and invertebrates. The TULP proteins share a conserved C-terminal region of approximately 200 amino acid residues.

Specifications

Antigen Cancer testis antigen 65
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TULP2
Gene Accession No. O00295
Gene Alias BOS_17630; cancer testis antigen 65; Cancer/testis antigen 65; CT65; Pdet; Protein P4-6; tubby like protein 2; Tubby-like protein 2; Tubby-related protein 2; TUBL2; Tulp2
Gene Symbols TULP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TULP2/Cancer testis antigen 65.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7288
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.