Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAND1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567031 Supplier Invitrogen™ Supplier No. PA567031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSIRLLALLSL GEVGHHIDLS GQLELKSVIL EAFSSPSEEV KSAASYALGS ISVGNLPEYL PFVLQEI Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

CAND1 enhances transcription from various types of promoters. It is a regulatory protein that interferes with the assembly of the SCF (SKP1-CUL1-F-box protein) ubiquitin ligase complex and thereby down-regulates ubiquitination of target proteins. CAND1 prevents neddylation of CUL1 by physically blocking access to the neddylation site. CAND1 disrupts interactions between CUL1 and SKP1A and between CUL1 and F-box proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAND1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAND1
Gene Accession No. Q86VP6
Gene Alias 2310038O07Rik; 6330512O03Rik; AI195005; AI846556; CAND1; cullin associated and neddylation disassociated 1; cullin associated and neddylation dissociated 1; cullin-associated and neddylation-dissociated 1; cullin-associated and neddylation-dissociated protein 1; Cullin-associated NEDD8-dissociated protein 1; D10Ertd516e; DKFZp434M1414; fb78a05; FLJ10114; FLJ10929; FLJ38691; FLJ90441; KIAA0829; mKIAA0829; p120 CAND1; TBP interacting protein; TBP-interacting protein; TBP-interacting protein 120A; TBP-interacting protein of 120 kDa A; TBP-interacting protein TIP120A; TIP120; TIP120 protein; TIP120A; Tp120a; wu:fb78a05; zgc:55729
Gene Symbols CAND1
Host Species Rabbit
Immunogen Recombinant Human CAND1. Recombinant protein control fragment (Product #RP-107671).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.