Learn More
CAND1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302900AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine and Orangutan
CAND1 enhances transcription from various types of promoters. It is a regulatory protein that interferes with the assembly of the SCF (SKP1-CUL1-F-box protein) ubiquitin ligase complex and thereby down-regulates ubiquitination of target proteins. CAND1 prevents neddylation of CUL1 by physically blocking access to the neddylation site. CAND1 disrupts interactions between CUL1 and SKP1A and between CUL1 and F-box proteins.Specifications
CAND1 | |
Polyclonal | |
Unconjugated | |
CAND1 | |
Cullin-associated and neddylation-dissociated 1, DKFZp434M1414, FLJ10114, FLJ10929, FLJ38691, FLJ90441, KIAA0829, TIP120, TIP120A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55832 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q86VP6 | |
CAND1 | |
Between 600 and 650 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.