Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAND2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120458
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120458 Supplier Invitrogen™ Supplier No. PA5120458
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, 293T, Jurkat, NIH/3T3, Mouse heart, Rat heart. The target is usually found in the following locations: Nucleus. Immunogen sequence: VGQVAGPGHQ RELKAVLLEA LGSPSEDVRA AASYALGRVG AGSLPDFLPF LLEQIEAEPR RQYLLLHSLR EALGAAQPDS LKPYAEDIWA LLFQRCEGAE EGTRGVVAEC IGKLVLVNPS FLLPRLRKQL AAGRPHTRST VITAVKFLIS D.

CAND2 may play a role in the assembly of ubiquitin ligase complexes and modulate the ubiquitination of target proteins. And it may be also a transcription regulator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAND2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CAND2
Gene Accession No. O75155, Q6ZQ73, Q9R0L4
Gene Alias 2210404G23Rik; cand1; Cand2; cullin associated and neddylation dissociated 2 (putative); cullin-associated and neddylation-dissociated 1; cullin-associated and neddylation-dissociated 2 (putative); cullin-associated and neddylation-dissociated protein 2; Cullin-associated NEDD8-dissociated protein 2; epididymis tissue protein Li 169; KIAA0667; mKIAA0667; p120 CAND2; si:ch211-257f16.2; si:dkey-72g22.1; TBP interacting protein; TBP-interacting protein; TBP-interacting protein 120B; TBP-interacting protein b; TBP-interacting protein of 120 kDa B; TBP-interacting protein Tip120B; Tip120B; Tp120b
Gene Symbols CAND2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192226, 23066, 67088
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.