Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAND2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543591
Description
Peptide sequence: MSTAAFHISS LLEKMTSSDK DFRFMATSDL MSELQKDSIQ LDEDSERKVV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
CAND2 may play a role in the assembly of ubiquitin ligase complexes and modulate the ubiquitination of target proteins. And it may be also a transcription regulator.
Specifications
CAND2 | |
Polyclonal | |
Unconjugated | |
CAND2 | |
2210404G23Rik; cand1; Cand2; cullin associated and neddylation dissociated 2 (putative); cullin-associated and neddylation-dissociated 1; cullin-associated and neddylation-dissociated 2 (putative); cullin-associated and neddylation-dissociated protein 2; Cullin-associated NEDD8-dissociated protein 2; epididymis tissue protein Li 169; KIAA0667; mKIAA0667; p120 CAND2; si:ch211-257f16.2; si:dkey-72g22.1; TBP interacting protein; TBP-interacting protein; TBP-interacting protein 120B; TBP-interacting protein b; TBP-interacting protein of 120 kDa B; TBP-interacting protein Tip120B; Tip120B; Tp120b | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23066 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75155 | |
CAND2 | |
Synthetic peptide directed towards the N-terminal of human CAND2 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction