Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAND2Rabbit anti-Human, Mouse, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501990097
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0097 Supplier Rockland Immunochemicals Supplier No. 100401A12
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This product reacts with human, rat and mouse CAND2/TIP120B. Cross reactivity does occur with human, rat and mouse CAND1/TIP120A. Cross reactivity with CAND2 from other sources is not known.

CAND2 may play a role in the assembly of ubiquitin ligase complexes and modulate the ubiquitination of target proteins. And it may be also a transcription regulator.

Specifications

Antigen CAND2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Formulation Antibody with 0.01% sodium azide; pH
Gene CAND2
Gene Accession No. O75155, Q6ZQ73, Q9R0L4
Gene Alias 2210404G23Rik; BOS_21118; cand1; Cand2; cullin associated and neddylation dissociated 2 (putative); cullin-associated and neddylation-dissociated 1; cullin-associated and neddylation-dissociated 2 (putative); Cullin-associated and neddylation-dissociated protein 2; Cullin-associated NEDD8-dissociated protein 2; Epididymis tissue protein Li 169; Kiaa0667; mKIAA0667; p120 CAND2; si:ch211-257f16.2; si:dkey-72g22.1; TBP interacting protein; TBP-interacting protein; TBP-interacting protein 120B; TBP-interacting protein b; TBP-interacting protein of 120 kDa B; TBP-interacting protein Tip120B; Tip120b; Tp120b
Gene Symbols CAND2
Host Species Rabbit
Immunogen This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 1130-1143 of Human CAND2/TIP120B (C-terminal) coupled to KLH.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192226, 23066, 67088
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.