Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Candesartan
SDP

Catalog No. 479150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-915-0 50 mg
47-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-915-0 Supplier Tocris Bioscience™ Supplier No. 4791/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AT1 receptor antagonist

Angiotensin II receptor 1 (AT1) antagonist (IC50 values are 1.12 and 2.86 nM in bovine adrenal cortex and rabbit aorta, respectively). Exhibits antihypertensive effects in animal models. Also available as a prodrug, candesartan cilexetil (Cat. No. 4792).

Chemical Identifiers

CAS 139481-59-7
Molecular Formula C24H20N6O3

Specifications

Quantity 50 mg
Formula Weight 440.45
Purity >98%
Molecular Weight (g/mol) 440.463
Chemical Name or Material Candesartan
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.