Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Candida albicans Recombinant Rabbit Monoclonal Antibody (6405)
GREENER_CHOICE

Catalog No. PIMA533304
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533304 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533304 Supplier Invitrogen™ Supplier No. MA533304
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Reactive with C. albicans (10 strains), C. parapsillosis, C. glabrata, C. tropicales & C. auris. Negative for C. krusei. Non-reactive with: Aspergillus species (4), Coccidioides, Histoplasma, Blastomyces, Paracoccidioides, Penicillium notatum, Mucor, Saccharomyces cerevisiae, and Pichia pastoris.

Candida albicans is the most frequently isolated fungal pathogen of humans, affecting immuocompromised patients ranging from premature infants to AIDS sufferers. Systemic infections have an attributed mortality of 30-50%. C. albicans is a diploid organism which has eight sets of homologous chromosomes. Its genome size is about 16 Mb (haploid), about 30% greater than S. cerevisiae (baker's yeast).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Candida albicans
Applications ELISA, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 6405
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.5
Gene Alias C. albicans
Host Species Rabbit
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.