Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Canine CCL2/MCP-1 Quantikine ELISA Kit
Click to view available options
Quantity:
1 x 96-well Plate
Description
The Canine CCL2/MCP-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies canine CCL2/JE/MCP-1 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Specifications
Specifications
Assay Range | 15.60 - 1000pg/mL |
Assay Sensitivity | 10.8pg/mL |
Conjugate | HRP |
Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
Specificity | Natural and Recombinant Canine MCP-1 |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 403981 |
Quantity | 1 x 96-well Plate |
Sample Volume | Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL) |
Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction