Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Canine CCL4/MIP-1 beta Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5839SP
This item is not returnable.
View return policy
Description
CCL4/MIP-1 beta Polyclonal specifically detects CCL4/MIP-1 beta in Canine samples. It is validated for Western Blot, Immunocytochemistry.Specifications
CCL4/MIP-1 beta | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, Exodus-3, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1 beta, MIP-1 beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2 | |
Sheep | |
Affinity Purified | |
RUO | |
6351 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
NP_001005250 | |
CCL4 | |
E. coli-derived recombinant canine CCL4/MIP-1 beta Ala24-Asn92 Accession # NP_001005250 | |
25 μg | |
Primary | |
Detects canine CCL4/MIP-1 beta in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human CCL4/MIP-1 beta and recombinant mouse CCL4/MIP-1 beta is observed and less than 1% cross-reactivity with recombinant viral CCL4/MIP-1 beta is observed. | |
Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction