Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine CCL5/RANTES Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6045SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6045SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6045SP Supplier R&D Systems Supplier No. MAB6045SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL5/RANTES Monoclonal specifically detects CCL5/RANTES in Canine samples. It is validated for Neutralization.

Specifications

Antigen CCL5/RANTES
Applications Neutralization
Classification Monoclonal
Clone 676117
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 0.075-0.45 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8HYS0
Gene Alias chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Gene Symbols CCL5
Host Species Mouse
Immunogen E. coli-derived recombinant canine CCL5/RANTES Ser24-Ser91 Accession # Q8HYS0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6352
Test Specificity Detects canine CCL5/RANTES in direct ELISAs. In Western blots, no cross-reactivity with recombinant cotton rat CCL5, recombinant feline CCL5, recombinant human (rh) CCL5, 17, 18, recombinant mouse (rm) CCL5, 7, or 18 is observed. In direct ELISAs, approximately 25% cross-reactivity with recombinant human CCL5 and no cross-reactivity with recombinant CCL5 from cotton rat, cat, or mouse is observed. No cross-reactivity with rhCCL1, 4, 11, 14, 16, 21, 23, 24, 26, 27, rmCCL6, 11, 21, 24, 26, or 27 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Canine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.