Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine CD25/IL-2R alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6227
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6227 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6227 Supplier R&D Systems Supplier No. AF6227
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CD25/IL-2R alpha Polyclonal specifically detects CD25/IL-2R alpha in Canine samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen CD25/IL-2R alpha
Applications Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Gene Accession No. BAI49682
Gene Alias CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Gene Symbols IL2RA
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant canine CD25/IL-2 R alpha Tyr19-Ile238 Accession # BAI49682
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3559
Test Specificity Detects canine CD25/IL-2 R alpha in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human CD25/IL-2 R alpha , recombinant mouse CD25/IL-2 R alpha , and recombinant rat CD25/IL-2 R alpha is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Canine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.