Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Canine CD34 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3346G100UG
This item is not returnable.
View return policy
Description
CD34 Monoclonal specifically detects CD34 in Canine samples. It is validated for Flow Cytometry.Specifications
CD34 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 | |
Mouse | |
100 μg | |
Primary | |
Detects canine CD34 in Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
1H6 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD34 | |
COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant canine CD34 | |
RUO | |
947 | |
Canine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction