Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine coronavirus Mouse anti-Virus, Clone: HM129L, Abnova™

Catalog No. 89035657 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-657 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-657 Supplier Abnova Corporation Supplier No. MAB13510
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against canine coronavirus.

  • High specificity
  • High reactivity

Specifications

Antigen Canine coronavirus
Applications ELISA
Classification Monoclonal
Clone HM129L
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against canine coronavirus.
Dilution ELISA The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Host Species Mouse
Purification Method Protein A purification
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with nucleoprotein of canine coronavirus.
Target Species Virus
Content And Storage Store at 4°C on dry atmosphere.
After reconstitution with 1 mL deionized water, store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.