Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Canine CXCL8/IL-8 ELISA Kit (Colorimetric)
SDP

Catalog No. NB599671KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB599671KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB599671KT Supplier Novus Biologicals™ Supplier No. NBP2599671KIT

High sensitivity and specificity for detection of dog IL-8.

The Canine CXCL8/IL-8 ELISA Kit (Colorimetric) measures canine CXCL8/IL-8 in serum, plasma, cell culture supernates, tissue homogenates.

Specifications

Assay Range 0.312 to 20ng/mL
Assay Sensitivity 0.078ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates
Specificity For detection of dog IL-8
Target Species Canine
For Use With (Application) ELISA
Gene ID (Entrez) 3576
Gene Symbol CXCL8, IL8
Kit Contents Assay plate (12 x 8 coated Microwells), Biotin-antibody Diluent, Biotin-antibody (100 x concentrate), Adhesive Strip (For 96 wells), HRP-avidin (100 x concentrate), HRP-avidin Diluent ,Wash Buffer (25 x concentrate), TMB Substrate, Standard (Freeze dried), Sample Diluent, Stop Solution, Instruction manual
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Immunology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
Target CXCL8/IL-8
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.