Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine Distemper Virus Antibody (7514), Novus Biologicals™
SDP

Catalog No. NB10062431 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062431 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062431 Supplier Novus Biologicals Supplier No. NB10062431

Mouse Monoclonal Antibody

Canine Distemper Virus Monoclonal specifically detects Canine Distemper Virus in Virus samples. It is validated for ELISA.

Specifications

Antigen Canine Distemper Virus
Applications ELISA
Classification Monoclonal
Clone 7514
Conjugate Unconjugated
Dilution ELISA 1:20-1:200
Gene Alias CDV
Host Species Mouse
Immunogen Envelope antigen of Canine distemper virus
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product detects Canine distemper virus (CDV), a negatively stranded RNA morbillivirus. CDV infection induces a systemic, often fatal disease in a broad and expanding host spectrum among the Carnivora. CDV is highly contagious, and transmission occurs predominantly via aerosols, resulting in neuroinvasiveness and severe immunosuppression. Mortality rates during CDV outbreaks can exceed 80%.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.