Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Canine Distemper Virus Monoclonal Antibody (DV2-12)
GREENER_CHOICE

Catalog No. PIMA182327
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA182327 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA182327 Supplier Invitrogen™ Supplier No. MA182327
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is morbillivirus specific and will detect phocine distemper virus, racoon distemper virus, and human measles virus, but does not cross-react with CAV2, CCV, CPI, or CPV. In Western blot, this antibody detects bands at ~76, 68, 54, and 32 kDa under reducing conditions. Heat-mediated antigen retrieval is recommended for the staining of paraffin sections. Mouse anti canine distemper virus antibody, clone DV2-12 recognizes canine distemper virus nucleoprotein. In western blots the antibody recognizes bands of approximately 76 kDa, 68 kDa, 54 kDa and 32 kDa under reducing conditions.

This antibody recognizes canine Distemper Virus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Canine Distemper Virus
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone DV2-12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide; pH 7.2
Gene Alias CDV
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Protein A
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.