Learn More
Invitrogen™ Canine/Feline Parvovirus Monoclonal Antibody (7353)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547585
Description
Specificity: Reactive with CPV and Feline Panleukopenia virus. Best Antibody Pairs for Antigen Detection (capture/conjugate): Canine/Feline Parvovirus Monoclonal antibody (Product # MA5-47585)/Canine Parvovirus Polyclonal antibody (Product # MA1-7467). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
CPV is reported to be a host range variant of FPV which emerged as a result of mutations of the viral capsid.
Specifications
Canine/Feline Parvovirus | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
7353 | |
Unconjugated | |
CPV; FPV; parvo | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.