Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine GM-CSF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1546
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1546 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1546 Supplier R&D Systems Supplier No. AF1546
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

GM-CSF Polyclonal specifically detects GM-CSF in Canine samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.

Specifications

Antigen GM-CSF
Applications Western Blot, Neutralization, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 2-8 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. P48749.1
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Gene Symbols CSF2
Host Species Goat
Immunogen E. coli-derived recombinant canine GM-CSF Ala18-Lys144 Accession # P48749.1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1437
Test Specificity Detects canine GM-CSF in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant rat GM-CSF is observed, approximately 5% cross-reactivity with recombinant feline GM-CSF and recombinant porcine GM-CSF is observed, and less than 1% cross-reactivity with recombinant human GM-CSF and recombinant mouse GM-CSF is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Canine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.