Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Canine Heartworm Monoclonal Antibody (A60Q)
GREENER_CHOICE

Catalog No. PIMA518177
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518177 Supplier Invitrogen™ Supplier No. MA518177
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not react with uninfected canine serum. Purity is >90%. Recommended pairs for ELISA: MA5-18178 (capture) and MA5-18177 (conjugated, detection).

Canine heartworm is a common parasite of dogs worldwide. Heartworm occurs in about 30 other species as well but rarely affects humans. The infection is commonly transmitted by mosquitoes. Adult heartworms can grow to be up to 10 inches in length and can live for 5-7 years in dogs affecting the heart and lungs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Canine Heartworm
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone A60Q
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Dirofilaria; dirofilariasis; Filaria
Host Species Mouse
Immunogen Canine Heartworm native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Nematode
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.