Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine heartworm Mouse anti-Parasite, Clone: FA306, Abnova™

Catalog No. 89033516 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-516 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-516 Supplier Abnova Corporation Supplier No. MAB13553
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against canine heartworm.

  • High specificity
  • High reactivity

Specifications

Antigen Canine heartworm
Classification Monoclonal
Clone FA306
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against canine heartworm.
Formulation In 50 mM PBS, pH 7.4 (0.09% sodium azide).
Host Species Mouse
Immunogen Native purified Dirofilaria immitis RG antigen.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Parasite
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.