Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Canine Heartworm Polyclonal Antibody, FITC
GREENER_CHOICE

Catalog No. PIPA173110
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173110 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173110 Supplier Invitrogen™ Supplier No. PA173110
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73110 antibody reacts with Dirofiliaria immitis. PA1-73110 has been successfully used in Immunofluorescence applications. The PA1-73110 antibody was raised against Dirofiliaria immitis adult worm preparation.

Canine heartworm is a common parasite of dogs worldwide. Heartworm occurs in about 30 other species as well but rarely affects humans. The infection is commonly transmitted by mosquitoes. Adult heartworms can grow to be up to 10 inches in length and can live for 5-7 years in dogs affecting the heart and lungs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Canine Heartworm
Applications Immunocytochemistry
Classification Polyclonal
Concentration 4-5 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Gene Alias Dirofilaria; dirofilariasis; Filaria
Host Species Goat
Immunogen Dirofiliaria immitis adult worm preparation.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Nematode
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.