Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine HGF Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3386
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3386 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3386 Supplier R&D Systems Supplier No. BAF3386
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

HGF Polyclonal specifically detects HGF in Canine samples. It is validated for Western Blot.

Specifications

Antigen HGF
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q867B7
Gene Alias deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A
Gene Symbols HGF
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant canine HGF Arg28-Arg494 (Lys266Gln, Trp321Arg, Leu458Pro) ( alpha chain) & Val495-Ser730 (Ile693Thr) ( beta chain) Accession # Q867B7
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3082
Test Specificity Detects canine HGF in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human (rh) HGF is observed, approximately 15% cross-reactivity with recombinant mouse HGF is observed, and less than 3% cross-reactivity with rhMSP is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Canine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.