Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Canine IFN-gamma Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems BAM7811
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Canine samples. It is validated for ELISA Detection (Matched Antibody Pair).Specifications
IFN-gamma | |
Monoclonal | |
LYOPH | |
ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
P42161 | |
IFNG | |
E. coli-derived recombinant canine IFN-gamma Gln24-Lys166 Accession # P42161 | |
250 μg | |
Primary | |
Detects canine IFN-gamma in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human, mouse, rat, cotton rat, feline, porcine, equine, or bovine IFN-gamma was observed. | |
Canine | |
Purified |
ELISA | |
142508 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3458 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction