Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine IL-1 beta/IL-1F2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3747
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3747 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3747 Supplier R&D Systems Supplier No. BAF3747
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-1 beta/IL-1F2 Polyclonal specifically detects IL-1 beta/IL-1F2 in Canine samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-1 beta/IL-1F2
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells
Gene Accession No. Q28292
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Gene Symbols IL1B
Host Species Goat
Immunogen E. coli-derived recombinant canine IL-1 beta/IL-1F2 Asp114-Ser265 Accession # Q28292
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3553
Test Specificity Detects canine IL-1 beta/IL-1F2 in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant feline IL-1 beta is observed, 10% cross-reactivity with recombinant human IL-1 beta , recombinant rhesus macaque IL-1 beta , recombinant equine IL-1 beta , and recombinant porcine IL-1 beta is observed, and 5% cross-reactivity with recombinant mouse IL-1 beta , recombinant rat IL-1 beta , and recombinant cotton rat IL-1 beta is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Canine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.