Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Canine IL-1 beta/IL-1F2 DuoSet ELISA
SDP

Catalog No. DY3747 Shop All Novus Biologicals Products
Change view
Click to view available options
Packaging Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DY3747 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY3747 Supplier R&D Systems™ Supplier No. DY3747

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)

Canine IL-1 beta/IL-1F2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Canine IL-1 beta/IL-1F2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies canine IL-1 beta/IL-1F2.

Specifications

Product Type ELISA Development Kit
Quantity 1 Kit
Species Canine
Conjugate Biotin
Assay Sensitivity 7.81 to 500 pg/mL
Assay Range 7.81 to 500 pg/mL
Cross Reactivity <50%cross-species reactivity observed with species tested
Format 96-well Strip Plate
For Use With (Application) ELISA
Gene ID (Entrez) 553, 16176, 24494, 397122, 403974, 100034237, 100135556
Product Line R&D Systems
Sample Type Cell Culture Supernatant
Sample Volume 100 μL
Synonym catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Kit Contents Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Packaging Quantity 15 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Cytokines
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.