Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine IL-10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF735
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF735 100 μg
AF735SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF735 Supplier R&D Systems Supplier No. AF735
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-10 Polyclonal specifically detects IL-10 in Canine samples. It is validated for Western Blot.

Specifications

Antigen IL-10
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. XP_855560
Gene Alias CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Gene Symbols IL10
Host Species Goat
Immunogen E. coli-derived recombinant canine IL-10 (R&D Systems, Catalog # 735-CL) Ser20-Ile179 Accession # XP_855560
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3586
Test Specificity Detects canine IL-10 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant feline IL-10, recombinant human IL-10, recombinant porcine IL-10 is observed and 5% cross-reactivity with recombinant mouse IL-10 and recombinant rat IL-10 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Canine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.