Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Canine IL-12/IL-23 p40 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1969
This item is not returnable.
View return policy
Description
IL-12/IL-23 p40 Polyclonal specifically detects IL-12/IL-23 p40 in Canine samples. It is validated for Western Blot.Specifications
IL-12/IL-23 p40 | |
Polyclonal | |
Biotin | |
Q28268 | |
IL12B | |
Mouse myeloma cell line NS0-derived recombinant canine IL-12/IL-23 p40 Ile23-Ser329 Accession # Q28268 | |
50 μg | |
Primary | |
Detects canine IL-12/IL-23 p40 in Western blots. | |
Canine | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40 | |
Goat | |
Affinity Purified | |
RUO | |
403976 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction