Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Canine IL-17 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5848SP
This item is not returnable.
View return policy
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Canine samples. It is validated for Neutralization.Specifications
IL-17/IL-17A | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3605 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Neutralization | |
665909 | |
Neutralization 5-30 ng/mL | |
NP_001159350 | |
IL17A | |
E. coli-derived recombinant canine IL-17 Gly26-Ala155 Accession # NP_001159350 | |
25 μg | |
Primary | |
Detects canine IL-17 in direct ELISAs. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) IL-17A is observed and no cross-reactivity with rhIL-17F or recombinant mouse IL-17A is observed. | |
Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction