Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine IL-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1815
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1815 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1815 Supplier R&D Systems Supplier No. AF1815
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-2 Polyclonal specifically detects IL-2 in Canine samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-2
Applications Western Blot, Neutralization, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.75-3.0 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. Q29416
Gene Alias aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF
Gene Symbols IL2
Host Species Goat
Immunogen E. coli-derived recombinant canine IL-2 (R&D Systems, Catalog # 1815-CL) Ala21-Thr155 with a Cys147Ser substitution Accession # Q29416
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3558
Test Specificity Detects canine IL-2 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human IL-2, recombinant mouse IL-2, recombinant rat IL-2, recombinant feline IL-2, recombinant cotton rat IL-2, recombinant bovine IL-2, recombinant equine IL-2, and recombinant porcine IL-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Canine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.