Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Canine IL-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB7541
This item is not returnable.
View return policy
Description
IL-4 Monoclonal specifically detects IL-4 in Canine samples. It is validated for Western Blot.Specifications
IL-4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3565 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
140417 | |
Western Blot 1 ug/mL | |
O77762 | |
IL4 | |
E. coli-derived recombinant canine IL-4 His25-His132 Accession # O77762 | |
500 μg | |
Primary | |
Detects recombinant canine IL-4 in direct ELISAs and Western blots. In direct ELISAs, 15% cross-reactivity with recombinant porcine IL-4 is observed, no cross-reactivity with recombinant human IL-13 is observed, and no cross-reactivity with IL-4 from human, cat, mouse, rat, or cotton rat is observed. | |
Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction