Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine IL-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF754
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF754 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF754 Supplier R&D Systems Supplier No. AF754
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-4 Polyclonal specifically detects IL-4 in Canine samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen IL-4
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 2-8 ug/mL
Gene Accession No. O77762
Gene Alias B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Gene Symbols IL4
Host Species Goat
Immunogen E. coli-derived recombinant canine IL-4 His25-His132 Accession # O77762
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3565
Test Specificity Detects canine IL-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human IL-4 is observed, 15% cross-reactivity with recombinant feline IL-4 and recombinant porcine IL-4 is observed and less than 1% cross-reactivity with recombinant cotton rat IL-4, recombinant mouse IL-4, and recombinant rat IL-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Canine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.