Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Canine KGF/FGF-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1957SP
This item is not returnable.
View return policy
Description
KGF/FGF-7 Monoclonal specifically detects KGF/FGF-7 in Canine samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
KGF/FGF-7 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P79150 | |
FGF7 | |
E. coli-derived recombinant canine KGF/FGF-7 Cys32-Thr194 Accession # P79150 | |
25 μg | |
Primary | |
Detects canine KGF/FGF-7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) FGF-11, -12, -13, -16, -20, -21, -22, -23, or rhFGFbasic is observed. | |
Canine | |
Purified |
Immunohistochemistry, Immunocytochemistry | |
299026 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2252 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction