Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

canine NKp46 (CD335) Mouse anti-Canine, Unconjugated, Clone: 48A, MilliporeSigma™

Catalog No. MABF21091MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF21091MI 100 μg
MABF21092MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF21091MI Supplier MilliporeSigma Supplier No. MABF2109100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

canine NKp46 (CD335) Monoclonal antibody specifically detects canine NKp46 (CD335) in Canine samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen canine NKp46 (CD335)
Applications ELISA, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 48A
Conjugate Unconjugated
Gene Accession No. NP_001271377
Gene Alias Natural cytotoxicity triggering receptor 1;NK cell-activating receptor;Natural killer cell p46-related protein;NK-p46;Canine NKp46;CD335
Gene Symbols NCR1
Host Species Mouse
Immunogen L-cells expressing canine NKp46 fusion protein
Molecular Weight of Antigen 36.13 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 48A is a mouse monoclonal antibody that specifically detects canine NKp46.
Target Species Canine
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.