Learn More
Canine Parvovirus Antibody (8H7), DyLight 550, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NB10073204R
Catalog No. NB10073204R
Only null left
Add to Cart
Description
Description
Canine Parvovirus Monoclonal specifically detects Canine Parvovirus in Virus samples. It is validated for ELISA, Sandwich ELISA.
Specifications
Specifications
Parvovirus
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Protein A purified
RUO
Parvovirus, Canine (8H7)
Store at 4C in the dark.
IgG2a
ELISA
8H7
ELISA, Sandwich ELISA
CPV, Parvo
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Canine parvovirus.
0.2 mL
Primary
Virus
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.