Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Canine Parvovirus Monoclonal Antibody (A92C)
GREENER_CHOICE

Catalog No. PIMA17467
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17467 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17467 Supplier Invitrogen™ Supplier No. MA17467
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7467 antibody reacts with Canine Parvovirus in viral and infected tissue samples. MA1-7467 has been successfully used in ELISA applications. The MA1-7467 antibody was raised against a parovirus viral preparation.

Canine parvovirus is a contagious virus mainly affecting dogs. CPV is highly contagious and is spread from dog to dog by direct or indirect contact with their feces. Vaccines can prevent this infection, but mortality can reach 91% in untreated cases. Treatment often involves veterinary hospitalization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Canine Parvovirus
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone A92C
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias CPV; parvo
Host Species Mouse
Immunogen Viral preparation.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.