Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine RAGE Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4750
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4750 100 μg
AF4750SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4750 Supplier R&D Systems Supplier No. AF4750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

AGER Polyclonal specifically detects AGER in Canine samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen RAGE
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. XP_532093
Gene Alias advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1
Gene Symbols MOK
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant canine RAGE Asp25-Val339 Accession # XP_532093
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5891
Test Specificity Detects canine RAGE in ELISAs and Western blots. In sandwich immunoassays, approximately 0.5% cross-reactivity with recombinant human RAGE is observed and less than 0.1% cross-reactivity with recombinant mouse RAGE and recombinant rat RAGE is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Canine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.