Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Canine TGF-beta 1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB601471KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB601471KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB601471KT Supplier Novus Biologicals™ Supplier No. NBP2601471KIT
Add to Cart
Add to Cart

High sensitivity and specificity for detection of dog TGF-B1.

The Canine TGF-beta 1 ELISA Kit (Colorimetric) measures canine TGF-beta 1 in serum, plasma.

Specifications

Assay Range 0.781 to 50ng/mL
Assay Sensitivity 0.747ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Specificity For detection of dog TGF-B1
Target Species Canine
For Use With (Application) ELISA
Gene ID (Entrez) 7040
Gene Symbol TGFB1
Kit Contents Instruction manual, Sample Diluent, Standard (Freeze dried), Stop Solution, Wash Buffer (25 x concentrate), TMB Substrate, HRP-avidin (100 x concentrate), HRP-avidin Diluent, Biotin-antibody (100 x concentrate), Biotin-antibody Diluent, Assay plate (12 x 8 coated Microwells), Adhesive Strip (For 96 wells), 1.2N NaOH/0.5M HEPES, 1 N HCI
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Signal Transduction
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Target TGF-beta 1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.