Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Canine TIM-1/KIM-1/HAVCR ELISA Kit (Colorimetric)
SDP

Catalog No. NB229074
Click to view available options
Quantity:
1 Kit

Specifications

Assay Range 156.1-10000 pg/mL
Assay Sensitivity 69 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Urine
Specificity Recognizes both recombinant and natural Canine TIM-1/KIM-1/HAVCR.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 26762
Gene Symbol HAVCR1
Quantity 1 Kit
Research Discipline CD Markers, Cellular Signaling, Immunology
Storage Requirements Storage is content dependent.
Synonym HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Target Canine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.