Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cannabinoid R2/CB2/CNR2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300606
Description
Cannabinoid R2/CB2/CNR2 Polyclonal specifically detects Cannabinoid R2/CB2/CNR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Cannabinoid R2/CB2/CNR2 | |
Polyclonal | |
Unconjugated | |
PBS, 1.0 mg/ml BSA and 30% Glycerol with 0.05% Sodium Azide | |
CNR2 | |
A fusion protein containing the first 33 amino acid residues from human CB2. | |
100 μL | |
GPCR, Neuroscience, Vision | |
1269 | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:250 - 1:2500, Flow Cytometry 1:50, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
cannabinoid receptor 2 (macrophage), CB-2, CB2cannabinoid receptor 2, CX5testis-dominant CNR2 isoform CB2, hCB2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Cannabinoid Receptor 2 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction