Learn More
Invitrogen™ Cannabinoid Receptor 1 Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Monkey, Marmoset, Mouse, Rat, Panda, Dog, Bat, Bovine, Cat, Horse, Pig (100%) Opossum (94%).
Specifications
Specifications
| Antigen | Cannabinoid Receptor 1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | CNR1 |
| Gene Accession No. | P20272, P21554, P47746 |
| Gene Alias | brain-type cannabinoid receptor; CANN6; Cannabinoid receptor 1; cannabinoid receptor 1 (brain); CB1; CB1 cannabinoid receptor; CB1 receptor; CB1A; CB1K5; CB1R; CB-R; central cannabinoid receptor; CNR; Cnr1; HGNC:2159; OTTHUMP00000016839; RP1-23D17.1; seven transmembrane domains; G-protein-coupled receptor; Skr6; SKR6R; striatal cannabinoid receptor type 1 protein |
| Gene Symbols | CNR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.