Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cannabinoid Receptor 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206351
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6351 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6351 Supplier Fabgennix Inc Supplier No. CB2R201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cannabinoid Receptor 2 Polyclonal antibody specifically detects Cannabinoid Receptor 2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Cannabinoid Receptor 2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene CNR2
Gene Accession No. P34972, P47936, Q9QZN9
Gene Alias Cannabinoid Receptor 2; cannabinoid receptor 2 (macrophage); CB2; CB-2; CB2 receptor; CB2A; CB2B; CB2C; CB2R; CB2-R; CNR2; CNR2C; CNR3; CX5; HCB2; HGNC:2160; OTTHUMP00000044841; peripheral cannabinoid receptor; rCB2; RP11-4M23.1; testis-dominant CNR2 isoform CB2
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 300-347 on mouse CB2 Receptor protein common to both CB2L and CB2S.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1269, 12802, 57302
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.