Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cannabinol Mouse anti-Human, Clone: 2B1, Bioss
SDP

Catalog No. 501979901
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9901 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9901 Supplier Bioss Supplier No. BSM2028M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cannabinol (CBN) is a weakly psychoactive cannabinoid found in Cannabis sativa and Cannabis indica. It is also a metabolite of tetrahydrocannabinol (THC). CBN acts as a weak agonist of CB1 receptors but has a higher affinity to CB2 receptors, with lower affinities in comparison to THC. Because of its somewhat selective CB2 agonism, it is used experimentally as an immunosuppressant.

Specifications

Antigen Cannabinol
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2B1
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Mouse
Immunogen KLH conjugated with Cannabinol.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.