Learn More
Stoelting™ Cannula Holder
Stoelting Cannula Holder was designed to hold 3.4 mm cannula heads, as manufactured by Plastics One™ for cannula sets, and DURECT for ALZET™ Osmotic Pumps
$855.00 - $1071.00
Specifications
For Use With (Application) | Neuroscience |
---|---|
For Use With (Equipment) | Cannulas |
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
10-000-381
![]() |
Stoelting™
516361 |
Cannula and Minipump Holder |
Each for $1,071.00
|
|
|||||
10-000-161
![]() |
Stoelting™
51636 |
Cannula holder |
Each for $855.00
|
|
|||||
Description
Non-intrusive for cementing cannulas in place. 51636-1 holds the cannula away from the shaft of the probe holder while the 51636 holds the cannula vertically directly in line with the shaft of the probe holder.Specifications
Neuroscience | |
Cannulas |