Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Canola Oil, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 g
2.5 kg
16 kg
Packaging:
Amber Glass Bottle
Poly Pail
Description
Spectrum™ Chemical Canola Oil
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Amber Glass Bottle |
| Quantity | 500 g |
| Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction