Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CANT1 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547864
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
This protein encoded by this gene belongs to the apyrase family. It functions as a calcium-dependent nucleotidase with a preference for UDP. Mutations in this gene are associated with Desbuquois dysplasia with hand anomalies.
Specifications
| CANT1 | |
| Polyclonal | |
| Unconjugated | |
| CANT1 | |
| 5830420C20Rik; Apy1h; apyrase 1, homolog (C. lectularius); apyrase homolog; Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase; calcium activated nucleotidase 1; Cant1; D11Bwg0554e; DBQD; DBQD1; EC 3.6.1.6; ectonucleoside triphosphate diphosphohydrolase 8; Entpd8; galectin-3-binding protein; soluble calcium-activated nucleotidase 1; lectin, galactoside-binding, soluble, 3 binding protein; LGALS3BP; micromelic dwarfism with vertebral and metaphyseal abnormalities and advanced carpotarsal ossification; putative MAPK-activating protein PM09; Putative NF-kappa-B-activating protein 107; SCAN1; SCAN-1; SHAPY; soluble Ca-activated nucleotidase, isozyme 1; soluble calcium-activated nucleotidase 1; soluble calcium-activated nucleotidase SCAN-1; srapy | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 124583 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8WVQ1 | |
| CANT1 | |
| Mouse myeloma cell line NS0-derived recombinant human Calcium Activated Nucleotidease 1/CANT1 Gly80-Ile401. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction