Learn More
Invitrogen™ CANT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592914
Description
Immunogen sequence: HRPAPGRPPT HNAHNWRLGQ APANWYNDTY PLSPPQRTPA GIRYRIAVIA DLDTESRAQE ENTWFSYLKK GYLTLSDSGD KVAVEWDKDH GVLESHLAEK GRGMELSDLI VFNGKLYSVD DRTGVVYQIE GSKAVPWVIL SDGDGTVEKG FKAEWLAVKD ERLYVGGLGK Positive Samples: U-251MG; Cellular Location: Endoplasmic reticulum membrane, Golgi apparatus, Golgi stack membrane, Single-pass type II membrane protein.
This protein encoded by this gene belongs to the apyrase family. It functions as a calcium-dependent nucleotidase with a preference for UDP. Mutations in this gene are associated with Desbuquois dysplasia with hand anomalies.
Specifications
CANT1 | |
Polyclonal | |
Unconjugated | |
CANT1 | |
5830420C20Rik; Apy1h; apyrase 1, homolog (C. lectularius); apyrase homolog; Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase; calcium activated nucleotidase 1; Cant1; D11Bwg0554e; DBQD; DBQD1; EC 3.6.1.6; ectonucleoside triphosphate diphosphohydrolase 8; Entpd8; galectin-3-binding protein; soluble calcium-activated nucleotidase 1; lectin, galactoside-binding, soluble, 3 binding protein; LGALS3BP; micromelic dwarfism with vertebral and metaphyseal abnormalities and advanced carpotarsal ossification; putative MAPK-activating protein PM09; Putative NF-kappa-B-activating protein 107; SCAN1; SCAN-1; SHAPY; soluble Ca-activated nucleotidase, isozyme 1; soluble calcium-activated nucleotidase 1; soluble calcium-activated nucleotidase SCAN-1; srapy | |
Rabbit | |
Affinity Chromatography | |
RUO | |
124583 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.84 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q8WVQ1 | |
CANT1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 62-231 of human CANT1 (NP_6201481). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.