Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cantharidin, 98%, Thermo Scientific Chemicals

Catalog No. aaj61801mc
Click to view available options
Quantity:
100 mg

 

Cantharidin is used as a potent inhibitor of PP2A phosphatase activity. It has been shown to stimulate cell cycle progression and induce premature mitosis, used topically (0.7%) as an anti-wart treatment, and has been shown to be active in cervical, tongue, neuroblastoma, bone, leukemia, ovarian, colon, and various other cancer cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cantharidin is used as a potent inhibitor of PP2A phosphatase activity. It has been shown to stimulate cell cycle progression and induce premature mitosis, used topically (0.7%) as an anti-wart treatment, and has been shown to be ac
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-25-7
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00134968
InChI Key DHZBEENLJMYSHQ-XCVPVQRUSA-N
Synonym cantharidin, cantharone, cantharidine, kantaridin, kantharidin, cantharides camphor, kantharidin german, caswell no. 157, 1,2-dimethyl-3,6-epoxyperhydrophthalic anhydride, unii-igl471wq8p
PubChem CID 5944
ChEBI CHEBI:64213
SMILES CC12C3CCC(C1(C(=O)OC2=O)C)O3

Specifications

Melting Point 215°C to 217°C
Color White
Quantity 100 mg
UN Number UN2811
Merck Index 14,1752
Solubility Information Soluble in DMSO (25mg/ml warm),acetone (8mg/ml),ethanol (8mg/ml warm),chloroform (1:65),ether (1:560),and ethyl acetate (1:150).
Formula Weight 196.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Cantharidin
Hazard Category H300-H315-H319-H335
Hazard Statement GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-263-3
RTECSNumber RN8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.