Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cantharidin 98.0+%, TCI America™
SDP

Catalog No. C0019100MG
Click to view available options
Quantity:
25 mg
100 mg

Chemical Identifiers

CAS 56-25-7
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00134968
InChI Key DHZBEENLJMYSHQ-XCVPVQRUSA-N
Synonym cantharidin, cantharone, cantharidine, kantaridin, kantharidin, cantharides camphor, kantharidin german, caswell no. 157, 1,2-dimethyl-3,6-epoxyperhydrophthalic anhydride, unii-igl471wq8p
PubChem CID 5944
ChEBI CHEBI:64213
SMILES CC12C3CCC(C1(C(=O)OC2=O)C)O3

Specifications

Melting Point 217°C
Color White-Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cantharidin
RTECSNumber RN8575000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.